2-Bromo-5-chlorothiophene ≥95.0%

Supplier: TCI America

B1996-5G B1996-25G
TCB1996-005GEA 115.34 CAD
TCB1996-005G TCB1996-25G
2-Bromo-5-chlorothiophene ≥95.0%
2-Bromo-5-chlorothiophene

CAS Number: 2873-18-9
MDL Number: MFCD00014523
Molecular Formula: C4H2BrClS
Molecular Weight: 197.47
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): 85
Specific Gravity (20/20): 1.80

Formula: C₄H₂BrClS
MW: 197.48 g/mol
Boiling Pt: 70 °C
Melting Pt: –2 °C
Density: 1.80 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00014523
CAS Number: 2873-18-9
EINECS: 220-708-5
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ef98e130106812d9c7b6a5d4ea85901cd1f5f13&vid=4ef98e130106812d9c7b6a5d4ea85901cd1f5f13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR