4-Bromo-2-chloroaniline ≥98.0%
Supplier: TCI America
|
Warning
|
B1993-25G
B1993-5G
TCB1993-025GEA
214.88
CAD
TCB1993-025G
TCB1993-005G
4-Bromo-2-chloroaniline ≥98.0%
4-Bromo-2-chloroaniline
CAS Number: 38762-41-3
MDL Number: MFCD00007660
Molecular Formula: C6H5BrClN
Molecular Weight: 206.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70
Formula:
C₆H₅BrClN Boiling Pt: 264 °C (1013 hPa) Melting Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007660 CAS Number: 38762-41-3 EINECS: 254-118-4 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...