Bis(diphenylphosphino)methane ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B1982-25G B1982-5G
TCB1982-25GEA 233.84 CAD
TCB1982-25G TCB1982-005G
Bis(diphenylphosphino)methane ≥97.0% (by GC, titration analysis)
Bis(diphenylphosphino)methane

CAS Number: 2071-20-7
MDL Number: MFCD00003537
Molecular Formula: C25H22P2
Molecular Weight: 384.40
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 120


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: Methylenebis(diphenylphosphine)

Formula: C₂₅H₂₂P₂
Melting Pt: 120 °C
MDL Number: MFCD00003537
CAS Number: 2071-20-7
EINECS: 218-194-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f2ef258089403d3cf4a33b694d7d75808b93ab3&vid=6f2ef258089403d3cf4a33b694d7d75808b93ab3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR