2-Bromo-1,4-dimethoxybenzene ≥97.0%

Supplier: TCI America
Warning

Synonyms: 1-Bromo-2,5-dimethoxybenzene

B1979-5G B1979-25G
TCB1979-005GEA 189.6 CAD
TCB1979-005G TCB1979-025G
2-Bromo-1,4-dimethoxybenzene ≥97.0%
2-Bromo-1,4-dimethoxybenzene

CAS Number: 25245-34-5
MDL Number: MFCD00008355
Molecular Formula: C8H9BrO2
Molecular Weight: 217.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 237
Flash Point (°C): 110
Specific Gravity (20/20): 1.51


Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 2-Bromo-1,4-dimethoxybenzene

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 237 °C
Density: 1.51 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008355
CAS Number: 25245-34-5
EINECS: 246-756-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9af2d9a1ff3a846881fbca36f49fc5b213e3e6c&vid=44fd86f799fd7d539b190cc00c0ff6ac8b4b7159&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR