(3,5-Bis(trifluoromethyl)phenyl)acetic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
B1976-5G
B1976-1G
TCB1976-005GEA
167.48
CAD
TCB1976-005G
TCB1976-001G
(3,5-Bis(trifluoromethyl)phenyl)acetic acid ≥97.0% (by titrimetric analysis)
(3,5-Bis(trifluoromethyl)phenyl)acetic acid
CAS Number: 85068-33-3
MDL Number: MFCD00009908
Molecular Formula: C10H6F6O2
Molecular Weight: 272.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 125
Formula:
(C₆H₃(CF₃)₂)CH₂COOH Boiling Pt: 276 °C (1013 hPa) Melting Pt: 125 °C |
MDL Number:
MFCD00009908 CAS Number: 85068-33-3 EINECS: 285-295-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...