(2-Bromophenyl)acetic acid ≥98.0%
Supplier: TCI America
Synonyms:
2-(2-Bromophenyl)acetic acid
B1957-5G
B1957-25G
TCB1957-005GEA
45.82
CAD
TCB1957-005G
TCB1957-025G
(2-Bromophenyl)acetic acid ≥98.0%
(2-Bromophenyl)acetic acid
CAS Number: 18698-97-0
MDL Number: MFCD00004314
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 106
Formula:
(C₆H₄Br)CH₂COOH Boiling Pt: 308 °C (1013 hPa) Melting Pt: 106 °C |
MDL Number:
MFCD00004314 CAS Number: 18698-97-0 EINECS: 242-509-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...