4-Bromo-α-methylbenzyl alcohol ≥99.0%

Supplier: TCI America

Synonyms: 4-Bromophenyl methyl carbinol, 1-(4-Bromophenyl)ethanol, p-Bromo-α-methylbenzyl alcohol

B1955-25G B1955-5G
TCB1955-025GEA 199.08 CAD
TCB1955-025G TCB1955-5G
4-Bromo-α-methylbenzyl alcohol ≥99.0%
4-Bromo-α-methylbenzyl alcohol

CAS Number: 5391-88-8
MDL Number: MFCD00004514
Molecular Formula: C8H9BrO
Molecular Weight: 201.06
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 134
Flash Point (°C): 63
Freezing point (°C): 34


Other Product Names / Synonyms: 1-(4-Bromophenyl)ethanol, 1-(4-Bromophenyl)ethyl Alcohol

Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 134 °C
Density: 1.469 g/cm³ (15.2 °C)
Flash Pt: 63 °C
MDL Number: MFCD00004514
CAS Number: 5391-88-8
EINECS: 226-389-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38992394dbef8a8959af0d969293e8c074bcbe4f&vid=38992394dbef8a8959af0d969293e8c074bcbe4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR