4-Bromo-2-fluorobenzoic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

B1950-25G B1950-5G
TCB1950-25GEA 303.36 CAD
TCB1950-25G TCB1950-005G
4-Bromo-2-fluorobenzoic acid ≥99.0% (by titrimetric analysis)
4-Bromo-2-fluorobenzoic acid

CAS Number: 112704-79-7
MDL Number: MFCD00143263
Molecular Formula: C7H4BrFO2
Molecular Weight: 219.01
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 212

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR