3',5'-Bis(trifluoromethyl)acetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Di(trifluormethyl)-5-ethanoylbenzol, 1-(3,5-Bis(trifluoromethyl)phenyl)ethanone, 1-[3,5-Bis(trifluoromethyl)phenyl]ethan-1-one

B1949-5G B1949-25G
TCB1949-005GEA 31.6 CAD
TCB1949-005G TCB1949-25G
3',5'-Bis(trifluoromethyl)acetophenone ≥98.0%
3',5'-Bis(trifluoromethyl)acetophenone

CAS Number: 30071-93-3
MDL Number: MFCD00009910
Molecular Formula: C10H6F6O
Molecular Weight: 256.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 82
Specific Gravity (20/20): 1.42

Formula: C₁₀H₆F₆O
MW: 256.15 g/mol
Boiling Pt: 85 °C
Density: 1.42 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00009910
CAS Number: 30071-93-3
EINECS: 250-023-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ba8f4fd67a07fd04941b6bd8b0de7256dff4b83&vid=3ba8f4fd67a07fd04941b6bd8b0de7256dff4b83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR