3-Bromo-5-chloro-2-hydroxybenzaldehyde ≥96.0%

Supplier: TCI America

B1948-25G B1948-5G
TCB1948-25GEA 586.81 CAD
TCB1948-25G TCB1948-005G
3-Bromo-5-chloro-2-hydroxybenzaldehyde ≥96.0%
3-Bromo-5-chloro-2-hydroxybenzaldehyde

CAS Number: 19652-32-5
MDL Number: MFCD00051690
Molecular Formula: C7H4BrClO2
Molecular Weight: 235.46
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 88


Other Product Names / Synonyms: 3-Bromo-5-chloro-2-hydroxybenzaldehyde

Formula: C₇H₄BrClO₂
Melting Pt: 88 °C
MDL Number: MFCD00051690
CAS Number: 19652-32-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=536dc1c85ef71e3d41509b5834ed74f8ab6dfa9e&vid=fecd1bedd07d71f39d94d3ac4e5f5c34d48b11d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR