(3-Bromophenyl)acetic acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America

B1927-25G B1927-5G
TCB1927-25GEA 317.58 CAD
TCB1927-25G TCB1927-005G
(3-Bromophenyl)acetic acid ≥95.0% (by GC, titration analysis)
(3-Bromophenyl)acetic acid

CAS Number: 1878-67-7
MDL Number: MFCD00004330
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 101

Formula: (C₆H₄Br)CH₂COOH
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 101 °C
MDL Number: MFCD00004330
CAS Number: 1878-67-7
EINECS: 217-522-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6da73bf687788749dc0eb621bfdcab6f09595e5a&vid=6da73bf687788749dc0eb621bfdcab6f09595e5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR