Methyl-2-bromo-5-nitrobenzoate ≥98.0%
Supplier: TCI America
B1916-25G
B1916-5G
TCB1916-25GEA
175.22
CAD
TCB1916-25G
TCB1916-005G
Methyl-2-bromo-5-nitrobenzoate ≥98.0%
Methyl 2-bromo-5-nitrobenzoate
CAS Number: 6942-36-5
MDL Number: MFCD00010867
Molecular Formula: C8H6BrNO4
Molecular Weight: 260.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82
Other Product Names / Synonyms: 2-Bromo-5-nitrobenzoic Acid Methyl Ester
Formula:
C₈H₆BrNO₄ Melting Pt: 82 °C |
MDL Number:
MFCD00010867 CAS Number: 6942-36-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...