Benzophenone imine ≥95.0%

Supplier: TCI America

Synonyms: Benzophenoneimine

B1912-5G B1912-25G
TCB1912-005GEA 66.36 CAD
TCB1912-005G TCB1912-025G
Benzophenone imine ≥95.0%
Benzophenone imine

CAS Number: 1013-88-3
MDL Number: MFCD00001760
Molecular Formula: C13H11N
Molecular Weight: 181.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 110
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: Diphenylmethanimine

Formula: C₁₃H₁₁N
Boiling Pt: 108 °C
Density: 1.08 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00001760
CAS Number: 1013-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdb262cc35a281e7af7e36850b0aa4e2c5575443&vid=fdb262cc35a281e7af7e36850b0aa4e2c5575443&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR