4-Bromo-2-methylanisole ≥98.0%

Supplier: TCI America

Synonyms: 5-Bromo-2-methoxytoluene, 1-Bromo-4-methoxy-3-methylbenzene, 4-Bromo-1-methoxy-2-methylbenzene

B1910-25G B1910-5G
TCB1910-025GEA 323.9 CAD
TCB1910-025G TCB1910-005G
4-Bromo-2-methylanisole ≥98.0%
4-Bromo-2-methylanisole

CAS Number: 14804-31-0
MDL Number: MFCD01321139
Molecular Formula: C8H9BrO
Molecular Weight: 201.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 124
Melting point (°C): 69


Other Product Names / Synonyms: 4-Bromo-2-methylanisole

Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 124 °C
Melting Pt: 69 °C
MDL Number: MFCD01321139
CAS Number: 14804-31-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR