4-Bromo-2,6-xylidine ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromo-2,6-dimethylaniline

B1909-25G B1909-100G
TCB1909-025GEA 109.02 CAD
TCB1909-025G TCB1909-100G
4-Bromo-2,6-xylidine ≥98.0%
4-Bromo-2,6-xylidine

CAS Number: 24596-19-8
MDL Number: MFCD00007826
Molecular Formula: C8H10BrN
Molecular Weight: 200.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 113


Other Product Names / Synonyms: 4-Bromo-2,6-xylidine

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 120
Melting Pt: 50
Flash Pt: 113 °C
MDL Number: MFCD00007826
CAS Number: 24596-19-8
EINECS: 246-337-9
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f9c739281cbc21d1e71030ca161d4d10f3a6c61&vid=4f9c739281cbc21d1e71030ca161d4d10f3a6c61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR