2-Bromo-4,6-xylidine ≥97.0%

Supplier: TCI America
Warning

Synonyms: 6-Bromo-2,4-dimethylaniline, 6-Bromo-2,4-xylidine, 2-Bromo-4,6-dimethylaniline

B1908-5G B1908-25G
TCB1908-005GEA 67.78 CAD
TCB1908-005G TCB1908-025G
2-Bromo-4,6-xylidine ≥97.0%
2-Bromo-4,6-xylidine

CAS Number: 41825-73-4
MDL Number: MFCD00047826
Molecular Formula: C8H10BrN
Molecular Weight: 200.08
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 45


Other Product Names / Synonyms: 6-Bromo-2,4-xylidine

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Melting Pt: 45 °C
MDL Number: MFCD00047826
CAS Number: 41825-73-4
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8a139817eaf200609e0bdd19acdbb479814aed2&vid=f8a139817eaf200609e0bdd19acdbb479814aed2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR