4-tert-Butylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-(tert-Butyl)aniline

B1900-25ML B1900-5ML
TCB1900-025MLEA 51.38 CAD
TCB1900-025ML TCB1900-005ML
4-tert-Butylaniline ≥98.0% (by GC, titration analysis)
4-tert-Butylaniline

CAS Number: 769-92-6
MDL Number: MFCD00007899
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 101
Freezing point (°C): 15
Specific Gravity (20/20): 0.95
Storage Temperature: 0-10°C

Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 230 °C
Melting Pt: 15 °C
Density: 0.95 (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007899
CAS Number: 769-92-6
EINECS: 212-215-9
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54b50bf8952f03b96ab1e2d12fbe2739a36aee09&vid=51da3aa9c2a48913b32d1f62e7252c87335dc969&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR