2-Bromo-3-methylpyridine ≥97.0%
Supplier: TCI America
|
Warning
|
B1894-5G
B1894-25G
TCB1894-005GEA
47.4
CAD
TCB1894-005G
TCB1894-025G
2-Bromo-3-methylpyridine ≥97.0%
2-Bromo-3-methylpyridine
CAS Number: 3430-17-9
MDL Number: MFCD00239380
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 219
Flash Point (°C): 113
Specific Gravity (20/20): 1.55
Other Product Names / Synonyms: 2-Bromo-3-picoline
Formula:
C₆H₆BrN Boiling Pt: 219 °C Density: 1.55 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00239380 CAS Number: 3430-17-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...