1-Bromo-4-propylbenzene ≥97.0%

Supplier: TCI America

B1889-5G B1889-25G
TCB1889-005GEA 55.3 CAD
TCB1889-005G TCB1889-25G
1-Bromo-4-propylbenzene ≥97.0%
1-Bromo-4-propylbenzene

CAS Number: 588-93-2
MDL Number: MFCD00012456
Molecular Formula: C9H11Br
Molecular Weight: 199.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 225
Flash Point (°C): 95
Specific Gravity (20/20): 1.29

Formula: C₉H₁₁Br
Boiling Pt: 225 °C
Melting Pt: –41.5 °C
Density: 1.29 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00012456
CAS Number: 588-93-2
EINECS: 454-790-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af3a9bac740be7924454314efd75c288d77d4499&vid=af3a9bac740be7924454314efd75c288d77d4499&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR