3'-Bromopropiophenone ≥98.0%

Supplier: TCI America

B1888-25G B1888-5G
TCB1888-25GEA 632 CAD
TCB1888-25G TCB1888-005G
3'-Bromopropiophenone ≥98.0%
3'-Bromopropiophenone

CAS Number: 19829-31-3
MDL Number: MFCD00000084
Molecular Formula: C9H9BrO
Molecular Weight: 213.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 41
Flash Point (°C): 110


Other Product Names / Synonyms: 3-Bromophenyl Ethyl Ketone

Formula: C₉H₉BrO
Boiling Pt: 145 °C
Melting Pt: 41 °C
Flash Pt: 110 °C
MDL Number: MFCD00000084
CAS Number: 19829-31-3
EINECS: 243-353-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a83a78862c38cd80c702584e97fa51718191f56&vid=9a83a78862c38cd80c702584e97fa51718191f56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR