3,5-Bis(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
3,5-Bis(trifluoromethyl)benzeneboronic acid, (3,5-Bis(trifluoromethyl)phenyl)metaboric acid, (3,5-Bis(trifluoromethyl)phenyl)boronic acid, Dihydroxy(3,5-bis(trifluoromethyl)phenyl)borane
B1886-1G
B1886-5G
B1886-25G
TCB1886-001GEA
34.76
CAD
TCB1886-001G
TCB1886-005G
TCB1886-25G
3,5-Bis(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
3,5-Bis(trifluoromethyl)phenylboronic acid
CAS Number: 73852-19-4
MDL Number: MFCD00051850
Molecular Formula: C8H5BF6O2
Molecular Weight: 257.93
Form: Crystal
Color: White
Melting point (°C): 220
Other Product Names / Synonyms: 3,5-Bis(trifluoromethyl)benzeneboronic Acid
Formula:
C₈H₅BF₆O₂ MW: 257.93 g/mol Melting Pt: 220 °C |
MDL Number:
MFCD00051850 CAS Number: 73852-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...