2-Formylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
2-Formylbenzeneboronic acid, (2-Formylphenyl)metaboric acid, 2-Boronobenzaldehyde, (2-Formylphenyl)boronic acid, Dihydroxy(2-formylphenyl)borane
B1873-5G
B1873-1G
TCB1873-005GEA
136.67
CAD
TCB1873-005G
TCB1873-001G
2-Formylphenylboronic acid (contains varying amounts of Anhydride)
2-Formylphenylboronic acid
CAS Number: 40138-16-7
MDL Number: MFCD00151822
Molecular Formula: C7H7BO3
Molecular Weight: 149.94
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 125
Other Product Names / Synonyms: Benzaldehyde-2-boronic Acid, 2-Formylbenzeneboronic Acid
Formula:
C₇H₇BO₃ MW: 149.94 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD00151822 CAS Number: 40138-16-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...