3,5-Bis(trifluoromethyl)phenol ≥96.0%

Supplier: TCI America

B1869-25G B1869-5G
TCB1869-025GEA 403.37 CAD
TCB1869-025G TCB1869-005G
3,5-Bis(trifluoromethyl)phenol ≥96.0%
3,5-Bis(trifluoromethyl)phenol

CAS Number: 349-58-6
MDL Number: MFCD00000386
Molecular Formula: C8H4F6O
Molecular Weight: 230.11
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 110
Freezing point (°C): 19
Specific Gravity (20/20): 1.53

Formula: C₈H₄F₆O
Boiling Pt: 88 °C
Melting Pt: 21 °C
Density: 1.53 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000386
CAS Number: 349-58-6
EINECS: 206-488-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89b8822ec15da2f3a8d936be224e617907d13afd&vid=89b8822ec15da2f3a8d936be224e617907d13afd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR