5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene ≥98.0%
Supplier: TCI America
Synonyms:
5-Iodo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Di(trifluormethyl)-5-iodbenzene, 1,3-Bis(trifluoromethyl)-5-iodobenzene, 1-Iodo-3,5-bis(trifluoromethyl)benzene, 3,5-Bis(trifluoromethyl)iodobenzene
CAS Number: 328-73-4
MDL Number: MFCD00040837
Molecular Formula: C8H3F6I
Molecular Weight: 340.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 74
Specific Gravity (20/20): 1.92
Other Product Names / Synonyms: 3,5-Bis(trifluoromethyl)iodobenzene
Formula:
C₈H₃F₆I Boiling Pt: 172 °C Density: 1.92 (20 °C) Flash Pt: 74 °C |
MDL Number:
MFCD00040837 CAS Number: 328-73-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...