1-Bromo-4-chloro-3-fluorobenzene ≥98.0%
Supplier: TCI America
|
Warning
|
B1861-25G
B1861-5G
TCB1861-25GEA
175.38
CAD
TCB1861-25G
TCB1861-005G
1-Bromo-4-chloro-3-fluorobenzene ≥98.0%
1-Bromo-4-chloro-3-fluorobenzene
CAS Number: 60811-18-9
MDL Number: MFCD00042519
Molecular Formula: C6H3BrClF
Molecular Weight: 209.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Specific Gravity (20/20): 1.73
Formula:
C₆H₃BrClF Boiling Pt: 142 °C Density: 1.73 (20 °C) |
MDL Number:
MFCD00042519 CAS Number: 60811-18-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...