4-Bromophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

B1858-25G B1858-1G B1858-5G
TCB1858-25GEA 227.52 CAD
TCB1858-25G TCB1858-001G TCB1858-005G
4-Bromophenylboronic acid (contains varying amounts of Anhydride)
4-Bromophenylboronic acid

CAS Number: 5467-74-3
MDL Number: MFCD00002104
Molecular Formula: C6H6BBrO2
Molecular Weight: 200.83
Form: Crystal
Color: White
Melting point (°C): 292


Other Product Names / Synonyms: 4-Bromobenzeneboronic Acid

Formula: C₆H₆BBrO₂
Melting Pt: 292 °C
Density: 1.67 g/cm³ (20 °C)
MDL Number: MFCD00002104
CAS Number: 5467-74-3
EINECS: 226-779-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d037b74423b4a23efbaa90db9c84801eeaec87c7&vid=d037b74423b4a23efbaa90db9c84801eeaec87c7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR