4-Bromo-2-fluoroanisole ≥98.0%
Supplier: TCI America
|
Warning
|
B1855-5G
B1855-25G
TCB1855-005GEA
53.72
CAD
TCB1855-005G
TCB1855-25G
4-Bromo-2-fluoroanisole ≥98.0%
4-Bromo-2-fluoroanisole
CAS Number: 2357-52-0
MDL Number: MFCD00011710
Molecular Formula: C7H6BrFO
Molecular Weight: 205.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 98
Specific Gravity (20/20): 1.60
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromo-3-fluoro-4-methoxybenzene
Formula:
C₇H₆BrFO Boiling Pt: 208 °C Melting Pt: 16 °C Density: 1.60 (20 °C) Flash Pt: 98 °C |
MDL Number:
MFCD00011710 CAS Number: 2357-52-0 EINECS: 219-096-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...