4-Bromo-3-nitrotoluene ≥96.0%

Supplier: TCI America
Warning

B1854-5G B1854-25G
TCB1854-005GEA 62.88 CAD
TCB1854-005G TCB1854-025G
4-Bromo-3-nitrotoluene ≥96.0%
4-Bromo-3-nitrotoluene

CAS Number: 5326-34-1
MDL Number: MFCD00024180
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 152
Flash Point (°C): 110
Freezing point (°C): 31

Formula: C₇H₆BrNO₂
Boiling Pt: 152 °C
Melting Pt: 35 °C
Density: 1.57 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00024180
CAS Number: 5326-34-1
EINECS: 226-203-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30ae5b2f341137eeb3abef347c3852bc00b272de&vid=b166cbdc803cc462cb72cd40d05645d360a8455a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR