2-Bromo-5-nitrotoluene ≥98.0%
Supplier: TCI America
B1853-5G
B1853-25G
TCB1853-005GEA
131.14
CAD
TCB1853-005G
TCB1853-025G
2-Bromo-5-nitrotoluene ≥98.0%
2-Bromo-5-nitrotoluene
CAS Number: 7149-70-4
MDL Number: MFCD00007281
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 143
Melting point (°C): 78
Formula:
C₇H₆BrNO₂ Boiling Pt: 143 °C Melting Pt: 78 °C |
MDL Number:
MFCD00007281 CAS Number: 7149-70-4 EINECS: 230-481-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...