4-Bromo-2-nitrotoluene ≥98.0%

Supplier: TCI America
Warning

B1843-5G B1843-25G
TCB1843-005GEA 26.86 CAD
TCB1843-005G TCB1843-025G
4-Bromo-2-nitrotoluene ≥98.0%
4-Bromo-2-nitrotoluene

CAS Number: 60956-26-5
MDL Number: MFCD00041243
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 46
Flash Point (°C): 110

Formula: C₇H₆BrNO₂
Boiling Pt: 256 °C
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD00041243
CAS Number: 60956-26-5
EINECS: 262-536-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83ecf00f9d0fef08a78228ededf159b02955309a&vid=a4eaed77561f84491b2a2e7b28ced9e52073b972&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR