2-Bromo-4-fluoroanisole ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Bromo-4-fluoro-1-methoxybenzene

B1830-5G B1830-25G
TCB1830-005GEA 47.4 CAD
TCB1830-005G TCB1830-025G
2-Bromo-4-fluoroanisole ≥98.0%
2-Bromo-4-fluoroanisole

CAS Number: 452-08-4
MDL Number: MFCD00012014
Molecular Formula: C7H6BrFO
Molecular Weight: 205.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Flash Point (°C): 98
Specific Gravity (20/20): 1.61


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromo-3-fluoro-6-methoxybenzene

Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 79 °C
Melting Pt: –27 °C
Density: 1.61 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00012014
CAS Number: 452-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9f0dfe5d474685e7bdf1646fa67ff5ed0e2f24e&vid=e9f0dfe5d474685e7bdf1646fa67ff5ed0e2f24e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR