2-Bromo-4-fluoroaniline 98,0

Supplier: TCI America
Warning

B1829-5G B1829-25G
TCB1829-005GEA 48.98 CAD
TCB1829-005G TCB1829-025G
2-Bromo-4-fluoroaniline 98,0
2-Bromo-4-fluoroaniline

CAS Number: 1003-98-1
MDL Number: MFCD00042462
Molecular Formula: C6H5BrFN
Molecular Weight: 190.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 221
Flash Point (°C): 104

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 221 °C
Melting Pt: 41
Density: 1.67 g/cm³ (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00042462
CAS Number: 1003-98-1
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a997892c616da8a790a8fa852262314b38408423&vid=a997892c616da8a790a8fa852262314b38408423&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR