6-Bromo-1-hexanol ≥95.0%

Supplier: TCI America
Warning

Synonyms: 6-Bromohexan-1-ol, Hexamethylene bromohydrin, 1-Bromo-6-hydroxyhexane

B1822-5G B1822-25G
TCB1822-005GEA 139.83 CAD
TCB1822-005G TCB1822-025G
6-Bromo-1-hexanol ≥95.0%
6-Bromo-1-hexanol

CAS Number: 4286-55-9
MDL Number: MFCD00002983
Molecular Formula: C6H13BrO
Molecular Weight: 181.07
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 114
Flash Point (°C): 110
Specific Gravity (20/20): 1.33
Storage Temperature: 0-10°C

Formula: BrH₂C(CH₂)₄CH₂OH
Boiling Pt: 114 °C
Density: 1.33 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002983
CAS Number: 4286-55-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91105080a1df94281774d72ca2656e496ebd9e54&vid=91105080a1df94281774d72ca2656e496ebd9e54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR