4-Bromo-2-chlorophenol ≥97.0%
Supplier: TCI America
B1819-25G
B1819-5G
TCB1819-025GEA
128.93
CAD
TCB1819-025G
TCB1819-005G
4-Bromo-2-chlorophenol ≥97.0%
4-Bromo-2-chlorophenol
CAS Number: 3964-56-5
MDL Number: MFCD00002166
Molecular Formula: C6H4BrClO
Molecular Weight: 207.45
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 234
Melting point (°C): 49
Flash Point (°C): 110
Formula:
C₆H₄BrClO Boiling Pt: 234 °C Melting Pt: 49 °C Density: 1.617 g/cm³ (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00002166 CAS Number: 3964-56-5 EINECS: 223-572-5 UN: 3082 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...