4-Bromo-2-furaldehyde ≥97.0%

Supplier: TCI America

B1816-5G B1816-1G
TCB1816-005GEA 1309.82 CAD
TCB1816-005G TCB1816-001G
4-Bromo-2-furaldehyde ≥97.0%
4-Bromo-2-furaldehyde

CAS Number: 21921-76-6
MDL Number: MFCD00671504
Molecular Formula: C5H3BrO2
Molecular Weight: 174.98
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 55
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Bromofurfural

Formula: C₅H₃BrO₂
Melting Pt: 55 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671504
CAS Number: 21921-76-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14b3a56408572238ed647e26d804860402751730&vid=14b3a56408572238ed647e26d804860402751730&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR