4-tert-Butylcalix[4]arene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: p-tert-Butylcalix[4]arene

B1809-5G B1809-1G B1809-25G
TCB1809-005GEA 255.96 CAD
TCB1809-005G TCB1809-001G TCB1809-25G
4-tert-Butylcalix[4]arene ≥98.0% (by HPLC)
4-tert-Butylcalix[4]arene

CAS Number: 60705-62-6
MDL Number: MFCD00066280
Molecular Formula: C44H56O4
Molecular Weight: 648.93
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: Tetra-tert-butyl(tetrahydroxy)calix[4]arene

Formula: C₄₄H₅₆O₄
MW: 648.93 g/mol
MDL Number: MFCD00066280
CAS Number: 60705-62-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8503525e18e98b533f816080bfda4655ec40f8ad&vid=8503525e18e98b533f816080bfda4655ec40f8ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR