Ethyl-3-bromobenzoate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 3-bromobenzoate, 3-Bromobenzoic acid ethyl ester

B1808-25G B1808-5G
TCB1808-025GEA 143.78 CAD
TCB1808-025G TCB1808-5G
Ethyl-3-bromobenzoate ≥98.0%
Ethyl 3-bromobenzoate

CAS Number: 24398-88-7
MDL Number: MFCD00013529
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 261
Flash Point (°C): 113
Specific Gravity (20/20): 1.44


Other Product Names / Synonyms: 3-Bromobenzoic Acid Ethyl Ester

Formula: C₉H₉BrO₂
Boiling Pt: 261 °C
Density: 1.44 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00013529
CAS Number: 24398-88-7
EINECS: 246-223-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a03ff471ddc4b77b0266c8f621fa793c6081220b&vid=a03ff471ddc4b77b0266c8f621fa793c6081220b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR