3-Bromo-5-iodobenzoic acid ≥98.0%

Supplier: TCI America
Warning

B1807-5G B1807-25G
TCB1807-005GEA 94.8 CAD
TCB1807-005G TCB1807-025G
3-Bromo-5-iodobenzoic acid ≥98.0%
3-Bromo-5-iodobenzoic acid

CAS Number: 188815-32-9
MDL Number: MFCD00191851
Molecular Formula: C7H4BrIO2
Molecular Weight: 326.92
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₇H₄BrIO₂
MDL Number: MFCD00191851
CAS Number: 188815-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed46a0ebebe5662b3fe0166f94969d9df325caba&vid=ed46a0ebebe5662b3fe0166f94969d9df325caba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR