1-Bromo-2,3-difluorobenzene ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2,3-Difluorobromobenzene
B1800-1G
B1800-5G
B1800-25G
TCB1800-001GEA
47.4
CAD
TCB1800-001G
TCB1800-005G
TCB1800-25G
1-Bromo-2,3-difluorobenzene ≥98.0%
1-Bromo-2,3-difluorobenzene
CAS Number: 38573-88-5
MDL Number: MFCD00061136
Molecular Formula: C6H3BrF2
Molecular Weight: 192.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 61
Specific Gravity (20/20): 1.71
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,3-Difluorobromobenzene
Formula:
C₆H₃BrF₂ MW: 192.99 g/mol Boiling Pt: 234 °C (765 mmHg) Density: 1.71 (20 °C) Flash Pt: 61 °C |
MDL Number:
MFCD00061136 CAS Number: 38573-88-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...