4-Bromoindole ≥97.0%

Supplier: TCI America

Synonyms: 4-Bromo-1H-indole

B1797-5G B1797-1G
TCB1797-005GEA 205.4 CAD
TCB1797-005G TCB1797-001G
4-Bromoindole ≥97.0%
4-Bromoindole

CAS Number: 52488-36-5
MDL Number: MFCD00671502
Molecular Formula: C8H6BrN
Molecular Weight: 196.05
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 110
Specific Gravity (20/20): 1.58

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 96 °C
Density: 1.58 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00671502
CAS Number: 52488-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f20ec4fcca7eb69689ba73311ca02e1f4eb69f5f&vid=f20ec4fcca7eb69689ba73311ca02e1f4eb69f5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR