6-Bromoindole ≥98.0%

Supplier: TCI America

B1794-5G B1794-1G
TCB1794-5GEA 284.24 CAD
TCB1794-5G TCB1794-001G
6-Bromoindole ≥98.0%
6-Bromoindole

CAS Number: 52415-29-9
MDL Number: MFCD00238550
Molecular Formula: C8H6BrN
Molecular Weight: 196.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 95
Flash Point (°C): 110

Formula: C₈H₆BrN
Melting Pt: 95 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00238550
CAS Number: 52415-29-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a882c056c2c40f66c487ce37fa8d9182f29ca58&vid=2a882c056c2c40f66c487ce37fa8d9182f29ca58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR