5-Bromo-m-xylene ≥98.0%

Supplier: TCI America
Warning

B1792-5G B1792-25G
TCB1792-005GEA 34.76 CAD
TCB1792-005G TCB1792-025G
5-Bromo-m-xylene ≥98.0%
5-Bromo-m-xylene

CAS Number: 556-96-7
MDL Number: MFCD00000087
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 87
Specific Gravity (20/20): 1.35


Other Product Names / Synonyms: 1-Bromo-3,5-dimethylbenzene

Formula: C₈H₉Br
Boiling Pt: 205 °C
Density: 1.35 (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000087
CAS Number: 556-96-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105e4a6fcec6564b4357f25c74e2a191d13b3c7b&vid=105e4a6fcec6564b4357f25c74e2a191d13b3c7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR