5-Bromo-o-anisaldehyde ≥98.0%

Supplier: TCI America

B1791-250G B1791-25G
TCB1791-250GEA 733.91 CAD
TCB1791-250G TCB1791-025G
5-Bromo-o-anisaldehyde ≥98.0%
5-Bromo-o-anisaldehyde

CAS Number: 25016-01-7
MDL Number: MFCD00003347
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 117


Other Product Names / Synonyms: 5-Bromo-2-methoxybenzaldehyde

Formula: C₈H₇BrO₂
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: 117 °C
MDL Number: MFCD00003347
CAS Number: 25016-01-7
EINECS: 246-564-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a8a4791a64a9b6113a55e21fe61482df7630f88&vid=79fd789cca91eb84752b97a051bfc9e53ff9b9c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR