3-Bromo-p-anisaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
3-Bromo-4-methoxybenzaldehyde
B1790-25G
B1790-5G
TCB1790-025GEA
145.36
CAD
TCB1790-025G
TCB1790-5G
3-Bromo-p-anisaldehyde ≥98.0%
3-Bromo-p-anisaldehyde
CAS Number: 34841-06-0
MDL Number: MFCD00016599
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 162
Melting point (°C): 50
Flash Point (°C): 110
Other Product Names / Synonyms: 3-Bromo-4-methoxybenzaldehyde
Formula:
C₈H₇BrO₂ MW: 215.05 g/mol Boiling Pt: 162 °C Melting Pt: 50 °C Flash Pt: 110 °C |
MDL Number:
MFCD00016599 CAS Number: 34841-06-0 EINECS: 252-241-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...