5-Bromo-2-fluorobenzaldehyde ≥95.0%

Supplier: TCI America
Warning

B1789-5G B1789-25G
TCB1789-005GEA 36.34 CAD
TCB1789-005G TCB1789-025G
5-Bromo-2-fluorobenzaldehyde ≥95.0%
5-Bromo-2-fluorobenzaldehyde

CAS Number: 93777-26-5
MDL Number: MFCD00070755
Molecular Formula: C7H4BrFO
Molecular Weight: 203.01
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Flash Point (°C): 109
Freezing point (°C): 23

Formula: C₇H₄BrFO
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: 49 °C
Density: 1.71 g/cm³ (20 °C)
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00070755
CAS Number: 93777-26-5
EINECS: 298-056-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a8cb90886707c13bc1c2f8ccfb02c0225d86421&vid=b4c1767e3a82af5470527d7c9d0c9d27e5b0d623&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR