(R)-(+)-4-Benzyl-2-oxazolidinone ≥98.0%
Supplier: TCI America
Synonyms:
(R)-4-Benzyl-2-oxazolidinone
B1786-25G
B1786-5G
TCB1786-025GEA
441.29
CAD
TCB1786-025G
TCB1786-005G
(R)-(+)-4-Benzyl-2-oxazolidinone ≥98.0%
(R)-(+)-4-Benzyl-2-oxazolidinone
CAS Number: 102029-44-7
MDL Number: MFCD00010846
Molecular Formula: C10H11NO2
Molecular Weight: 177.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 87
Specific rotation [a]20/D: 14.5 deg (C=5, MeOH)
Formula:
C₁₀H₁₁NO₂ Melting Pt: 87 °C |
MDL Number:
MFCD00010846 CAS Number: 102029-44-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...