1-Bromo-4-(trifluoromethoxy)benzene ≥98.0%

Supplier: TCI America

Synonyms: 4-(Trifluoromethoxy)bromobenzene

B1772-5G B1772-25G
TCB1772-005GEA 33.18 CAD
TCB1772-005G TCB1772-025G
1-Bromo-4-(trifluoromethoxy)benzene ≥98.0%
1-Bromo-4-(trifluoromethoxy)benzene

CAS Number: 407-14-7
MDL Number: MFCD00040834
Molecular Formula: C7H4BrF3O
Molecular Weight: 241.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 67
Specific Gravity (20/20): 1.64

Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
Boiling Pt: 80 °C
Density: 1.64 (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00040834
CAS Number: 407-14-7
EINECS: 206-979-2
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=001c6bd25452f00a43fe7729faf8c71913aa793e&vid=44aff4b1f57b499a9c9c0ab50006beca44317fd1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR