2-Butylaniline ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1766-25G B1766-5G
TCB1766-25GEA 984.34 CAD
TCB1766-25G TCB1766-5G
2-Butylaniline ≥98.0% (by titrimetric analysis)
2-Butylaniline

CAS Number: 2696-85-7
MDL Number: MFCD00130033
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 126
Specific Gravity (20/20): 0.95

Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 126 °C
Density: 0.95 (20 °C)
MDL Number: MFCD00130033
CAS Number: 2696-85-7
EINECS: 220-272-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64bee1670c5d8d513e2fabf15274f64b9b44637a&vid=64bee1670c5d8d513e2fabf15274f64b9b44637a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR