(S)-(-)-4-Benzyl-2-oxazolidinone ≥98.0%
Supplier: TCI America
B1754-5G
B1754-25G
TCB1754-005GEA
150.1
CAD
TCB1754-005G
TCB1754-025G
(S)-(-)-4-Benzyl-2-oxazolidinone ≥98.0%
(S)-(-)-4-Benzyl-2-oxazolidinone
CAS Number: 90719-32-7
MDL Number: MFCD00064496
Molecular Formula: C10H11NO2
Molecular Weight: 177.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 88
Specific rotation [a]20/D: -14.5 deg (C=5, MeOH)
Formula:
C₁₀H₁₁NO₂ Melting Pt: 88 °C |
MDL Number:
MFCD00064496 CAS Number: 90719-32-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...