1-Bromo-3,5-bis(trifluoromethyl)benzene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 3,5-Bis(trifluoromethyl)-1-bromobenzene, 1-Bromo-3,5-bis(trifluoromethyl)benzene, 5-Bromo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Bis(trifluoromethyl)-5-bromobenzene, 3,5-Bis(trifluoromethyl)bromobenzene, 5-Bromo-α,α,α,α',α',α'-Hexafluoro-m-xylene

B1753-10G B1753-25G
TCB1753-010GEA 26.86 CAD
TCB1753-010G TCB1753-025G
1-Bromo-3,5-bis(trifluoromethyl)benzene ≥98.0%
1-Bromo-3,5-bis(trifluoromethyl)benzene

CAS Number: 328-70-1
MDL Number: MFCD00000381
Molecular Formula: C8H3BrF6
Molecular Weight: 293.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Flash Point (°C): 110
Specific Gravity (20/20): 1.73


Other Product Names / Synonyms: 3,5-Bis(trifluoromethyl)bromobenzene

Formula: C₈H₃BrF₆
Boiling Pt: 151 °C
Melting Pt: –16 °C
Density: 1.73 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000381
CAS Number: 328-70-1
EINECS: 206-334-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de30a43f490cea74eef7eeae6df7be500e33b033&vid=de30a43f490cea74eef7eeae6df7be500e33b033&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR